Product Details

SNP ID
rs185898822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4821609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGCCTCCATAAGCCTATGTACT[A/G]TTTCCTCTCTATGCTTTCAGCCACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51F2 PubMed Links

Gene Details

Gene
OR51F2
Gene Name
olfactory receptor family 51 subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004753.1 224 Missense Mutation TAT,TGT Y75C NP_001004753.1

View Full Product Details