Product Details

SNP ID
rs185682627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58516414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACCCGAGTGGATGAACATGTG[C/T]TTGGTGTAGTTTTTCCGGGAGCTGA
Phenotype
MIM: 603314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC27A5 PubMed Links

Gene Details

Gene
SLC27A5
Gene Name
solute carrier family 27 member 5
There are no transcripts associated with this gene.

Gene
ZBTB45
Gene Name
zinc finger and BTB domain containing 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316978.1 1571 Silent Mutation AAA,AAG K420K NP_001303907.1
NM_001316979.1 1571 Silent Mutation AAA,AAG K420K NP_001303908.1
NM_001316980.1 1571 Silent Mutation AAA,AAG K420K NP_001303909.1
NM_001316981.1 1571 Silent Mutation AAA,AAG K420K NP_001303910.1
NM_001316982.1 1571 Silent Mutation AAA,AAG K420K NP_001303911.1
NM_032792.3 1571 Silent Mutation AAA,AAG K420K NP_116181.1
XM_006723445.3 1571 Silent Mutation AAA,AAG K420K XP_006723508.1

View Full Product Details