Product Details

SNP ID
rs185687051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56113474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTTTCCCAAACTTAACTGTCCT[C/G]CAGTGCGGCAGTGAGGCTCCCCTGC
Phenotype
MIM: 190060
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MOS PubMed Links

Gene Details

Gene
MOS
Gene Name
v-mos Moloney murine sarcoma viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005372.1 509 Missense Mutation GCA,GGA A170G NP_005363.1

View Full Product Details