Product Details

SNP ID
rs186258915
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174349123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTCTGCTCTTCTGAAACTTTT[C/T]CTTTTTTCTATCTTTTTTCTTTTTT
Phenotype
MIM: 605228
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CIR1 PubMed Links

Gene Details

Gene
CIR1
Gene Name
corepressor interacting with RBPJ, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004882.3 Intron NP_004873.3

View Full Product Details