Product Details

SNP ID
rs186549879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:110476993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATCTCCTGACCATGGAGTTCGC[C/T]GGCTGGCTGAACGGCTCCCTCGTGG
Phenotype
MIM: 138297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENPEP PubMed Links

Gene Details

Gene
ENPEP
Gene Name
glutamyl aminopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001977.3 921 Silent Mutation GCC,GCT A193A NP_001968.3
XM_017007877.1 921 Intron XP_016863366.1

View Full Product Details