Product Details

SNP ID
rs186563897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34225491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGATCAGGAAATGTGGGATAAG[A/G]AGTATCGGAGGTTGCAGCTTTAGTT
Phenotype
MIM: 616245 MIM: 604878
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC4 PubMed Links

Gene Details

Gene
EMC4
Gene Name
ER membrane protein complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286420.1 Intron NP_001273349.1
NM_016454.3 Intron NP_057538.1
Gene
SLC12A6
Gene Name
solute carrier family 12 member 6
There are no transcripts associated with this gene.

View Full Product Details