Product Details

SNP ID
rs186629223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48596415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCCTCCCTTTCCAGCACCTTC[A/C]CTCGGCCTGTTTTTCTTCCTCCTCC
Phenotype
MIM: 142966 MIM: 142960 MIM: 142961
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOXB-AS3 PubMed Links

Gene Details

Gene
HOXB-AS3
Gene Name
HOXB cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXB3
Gene Name
homeobox B3
There are no transcripts associated with this gene.

Gene
HOXB5
Gene Name
homeobox B5
There are no transcripts associated with this gene.

Gene
HOXB6
Gene Name
homeobox B6
There are no transcripts associated with this gene.

View Full Product Details