Product Details
- SNP ID
-
rs186625503
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:65526285 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGACCTCCCCGCAAGGGGATCCC[C/T]GAGCTTGAGCAGTATGACCCCCCGG
- Phenotype
-
MIM: 607982
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCYL1
PubMed Links
Gene Details
- Gene
- SCYL1
- Gene Name
- SCY1 like pseudokinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001048218.1 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
NP_001041683.1 |
NM_020680.3 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
NP_065731.3 |
XM_005274118.3 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
XP_005274175.1 |
XM_005274120.3 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
XP_005274177.1 |
XM_005274121.3 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
XP_005274178.1 |
XM_017018029.1 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
XP_016873518.1 |
XM_017018030.1 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
XP_016873519.1 |
XM_017018031.1 |
614 |
Silent Mutation |
CCC,CCT |
P179P |
XP_016873520.1 |
XM_017018032.1 |
614 |
Silent Mutation |
CCC,CCT |
P36P |
XP_016873521.1 |
XM_017018033.1 |
614 |
Silent Mutation |
CCC,CCT |
P36P |
XP_016873522.1 |
View Full Product Details