Product Details

SNP ID
rs186327050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17203081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGCATGTAGCCTGGGTCTTGAT[C/G]GTGCCGGACCTGGATCCCTGCAGAA
Phenotype
MIM: 607273 MIM: 614960
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLCN PubMed Links

Gene Details

Gene
FLCN
Gene Name
folliculin
There are no transcripts associated with this gene.

Gene
PLD6
Gene Name
phospholipase D family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178836.3 491 Missense Mutation CAT,GAT H149D NP_849158.2
XM_017024310.1 491 Silent Mutation ACC,ACG T147T XP_016879799.1

View Full Product Details