Product Details

SNP ID
rs186352953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47370724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTGCCCTGATTCCAGGAGAAC[C/T]TGGCAGATCTTTTGAGGTAAGGAGG
Phenotype
MIM: 300024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF157 PubMed Links

Gene Details

Gene
ZNF157
Gene Name
zinc finger protein 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003446.3 125 Missense Mutation CCT,CTT P19L NP_003437.2

View Full Product Details