Product Details

SNP ID
rs186367234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128457328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTACTGACCCTACTCTCCATCTT[C/T]GTTAGAGTGATGGAGTCCCTAGAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HILPDA PubMed Links

Gene Details

Gene
HILPDA
Gene Name
hypoxia inducible lipid droplet associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098786.1 208 Silent Mutation TTC,TTT F20F NP_001092256.1
NM_013332.3 208 Silent Mutation TTC,TTT F20F NP_037464.1

View Full Product Details