Product Details

SNP ID
rs186432230
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:153969366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCCAGCATTTATGGTGCCTGAT[G/T]CCTGCATGGTCAGTGAGCTGCCCTT
Phenotype
MIM: 607138 MIM: 604671 MIM: 604740
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001255978.1 692 Missense Mutation GCC,TCC A152S NP_001242907.1
NM_001255979.1 692 Missense Mutation GCC,TCC A152S NP_001242908.1
NM_001255980.1 692 Missense Mutation GCC,TCC A132S NP_001242909.1
NM_001255981.1 692 Missense Mutation GCC,TCC A132S NP_001242910.1
NM_130898.3 692 Missense Mutation GCC,TCC A152S NP_570968.1
XM_006711172.2 692 Missense Mutation GCC,TCC A132S XP_006711235.1
XM_017000372.1 692 Missense Mutation GCC,TCC A132S XP_016855861.1
Gene
JTB
Gene Name
jumping translocation breakpoint
There are no transcripts associated with this gene.

Gene
MIR6737
Gene Name
microRNA 6737
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details