Product Details

SNP ID
rs186419762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29044910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACCCACATAGCTGATGGTCTTA[C/T]CAGGTCCCCACAAGTTGACCAGCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100129636 PubMed Links

Gene Details

Gene
LOC100129636
Gene Name
uncharacterized LOC100129636
There are no transcripts associated with this gene.

Gene
OR2W1
Gene Name
olfactory receptor family 2 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030903.3 266 Missense Mutation GAT,GGT D89G NP_112165.1

View Full Product Details