Product Details
- SNP ID
-
rs186180373
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:6026851 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAGGAGGAATAAGGTGATGAAGG[A/G]CGTTCAGCAGGATCAGGATGTCCAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR56A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11040335] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR56A1
- Gene Name
- olfactory receptor family 56 subfamily A member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001917.2 |
891 |
Missense Mutation |
GCC,GTC |
A285V |
NP_001001917.2 |
View Full Product Details