Product Details

SNP ID
rs186199942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74073599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTCCGCATCAACGTAGTCCAC[A/G]TTGTATGTCTGTCCCAAGAAGCTAG
Phenotype
MIM: 609093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO15 PubMed Links

Gene Details

Gene
FBXO15
Gene Name
F-box protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142958.1 1485 Silent Mutation AAC,AAT N465N NP_001136430.1
NM_152676.2 1485 Silent Mutation AAC,AAT N389N NP_689889.1
XM_011525856.1 1485 Silent Mutation AAC,AAT N395N XP_011524158.1
XM_011525858.2 1485 Intron XP_011524160.1
XM_011525859.1 1485 Intron XP_011524161.1
XM_011525860.1 1485 Intron XP_011524162.1
XM_017025603.1 1485 Silent Mutation AAC,AAT N389N XP_016881092.1
XM_017025604.1 1485 Silent Mutation AAC,AAT N389N XP_016881093.1

View Full Product Details