Product Details

SNP ID
rs186816860
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:152720012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGTGTCACTGACCCATGCTCTG[C/T]TCCCTGTTCCACCAGCTACTGCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf68 PubMed Links

Gene Details

Gene
C1orf68
Gene Name
chromosome 1 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024679.2 491 Missense Mutation GCT,GTT A164V NP_001019850.1

View Full Product Details