Product Details
- SNP ID
-
rs187146043
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:46230303 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCATGACTCCTTACCTGATGCTG[G/T]GCCAGCCACACAAGAGATGTGGGGA
- Phenotype
-
MIM: 610544
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IGFL1
PubMed Links
Gene Details
- Gene
- IGFL1
- Gene Name
- IGF like family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198541.1 |
132 |
Missense Mutation |
GGC,TGC |
G37C |
NP_940943.1 |
View Full Product Details