Product Details

SNP ID
rs187167681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:33437578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGCCCTATCCCTGTTTGAGAT[G/T]GGGATGTTTAGAATCTGTCCCAAGA
Phenotype
MIM: 609531
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RASGRP3 PubMed Links

Gene Details

Gene
RASGRP3
Gene Name
RAS guanyl releasing protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139488.1 Intron NP_001132960.1
NM_015376.2 Intron NP_056191.1
NM_170672.2 Intron NP_733772.1
XM_005264246.3 Intron XP_005264303.1
XM_011532746.2 Intron XP_011531048.1
XM_011532747.2 Intron XP_011531049.1
XM_011532748.2 Intron XP_011531050.2
XM_017003759.1 Intron XP_016859248.1
XM_017003760.1 Intron XP_016859249.1
XM_017003761.1 Intron XP_016859250.1
XM_017003762.1 Intron XP_016859251.1
XM_017003763.1 Intron XP_016859252.1

View Full Product Details