Product Details

SNP ID
rs187128979
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74415598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCCACGACCTTCCTAAAGGGAT[A/C]CGGTCCACGCCCCCAGGGTCCTGGG
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145054.1 649 Silent Mutation CGG,CGT R166R NP_001138526.1
NM_001316764.1 649 Silent Mutation CGG,CGT R193R NP_001303693.1
NM_001316765.1 649 Silent Mutation CGG,CGT R144R NP_001303694.1
NM_001316766.1 649 Silent Mutation CGG,CGT R98R NP_001303695.1
Gene
WDR54
Gene Name
WD repeat domain 54
There are no transcripts associated with this gene.

View Full Product Details