Product Details

SNP ID
rs187124967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144004902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCAAGGAAGCTCTGAAGAAACT[G/T]GCATATTGCCAGGCCAGCAGATCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6B1 PubMed Links

Gene Details

Gene
OR6B1
Gene Name
olfactory receptor family 6 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005281.1 906 Silent Mutation CTG,CTT L302L NP_001005281.1

View Full Product Details