Product Details

SNP ID
hCV182494060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144053381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGACCTTGCCTGGCGCCCGCAGG[A/G]CCTCCGTCACCGCTGGATGGACAGT
Phenotype
MIM: 614243 MIM: 610874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6846 PubMed Links

Gene Details

Gene
MIR6846
Gene Name
microRNA 6846
There are no transcripts associated with this gene.

Gene
OPLAH
Gene Name
5-oxoprolinase (ATP-hydrolysing)
There are no transcripts associated with this gene.

Gene
SMPD5
Gene Name
sphingomyelin phosphodiesterase 5
There are no transcripts associated with this gene.

Gene
SPATC1
Gene Name
spermatogenesis and centriole associated 1
There are no transcripts associated with this gene.

View Full Product Details