Product Details

SNP ID
rs186878328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37232139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTTGGAGAGGGAAATACTTCA[C/G]CTCCGGGAATGAGCCTGCAGAAAAC
Phenotype
MIM: 612828 MIM: 615898
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEBPZ PubMed Links

Gene Details

Gene
CEBPZ
Gene Name
CCAAT/enhancer binding protein zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005760.2 164 Intron NP_005751.2
Gene
NDUFAF7
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083946.1 164 Missense Mutation ACC,AGC T30S NP_001077415.1
NM_144736.4 164 Missense Mutation ACC,AGC T30S NP_653337.1
XM_005264410.1 164 UTR 5 XP_005264467.1
XM_005264411.1 164 Missense Mutation ACC,AGC T30S XP_005264468.1
XM_011532956.1 164 UTR 5 XP_011531258.1

View Full Product Details