Product Details

SNP ID
rs186885985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136497802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCGATGCCGGCGGAGAACCCGAT[G/T]CTCTCGGGCAGCAGCCGACGGACAC
Phenotype
MIM: 300107 MIM: 300346
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BRS3 PubMed Links

Gene Details

Gene
BRS3
Gene Name
bombesin receptor subtype 3
There are no transcripts associated with this gene.

Gene
HTATSF1
Gene Name
HIV-1 Tat specific factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163280.1 540 Missense Mutation GCT,TCT A40S NP_001156752.1
NM_014500.4 540 Missense Mutation GCT,TCT A40S NP_055315.2
XM_005262404.3 540 Missense Mutation GCT,TCT A40S XP_005262461.1

View Full Product Details