Product Details

SNP ID
rs186871647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30250720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTGCTGCTGCAGCTGTGTCAT[G/T]TACCGAATCTGACGAAGGTGCCAAA
Phenotype
MIM: 300097 MIM: 300153
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAGEB1 PubMed Links

Gene Details

Gene
MAGEB1
Gene Name
MAGE family member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002363.4 390 Missense Mutation TGT,TTT C76F NP_002354.2
NM_177404.2 390 Missense Mutation TGT,TTT C76F NP_796379.1
NM_177415.2 390 Missense Mutation TGT,TTT C76F NP_803134.1
Gene
MAGEB4
Gene Name
MAGE family member B4
There are no transcripts associated with this gene.

View Full Product Details