Product Details

SNP ID
rs186850947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33499220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCAGAAACCTGCTGCACTTTTC[C/T]GATATAGTTCACCACTTTTCTGTCT
Phenotype
MIM: 614426
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAK16 PubMed Links

Gene Details

Gene
MAK16
Gene Name
MAK16 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032509.3 2006 UTR 3 NP_115898.2
Gene
TTI2
Gene Name
TELO2 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102401.2 2006 Missense Mutation AGA,GGA R494G NP_001095871.1
NM_001265581.1 2006 Missense Mutation AGA,GGA R494G NP_001252510.1
NM_025115.3 2006 Missense Mutation AGA,GGA R494G NP_079391.2
XM_011544665.2 2006 Missense Mutation AGA,GGA R463G XP_011542967.1

View Full Product Details