Product Details

SNP ID
rs186937839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65385086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCCTGCCCAGGAACCAGCCCA[C/G]CTCAGCCCGCTTGGTTGAGCCCAGG
Phenotype
MIM: 604184 MIM: 616810
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IGDCC3 PubMed Links

Gene Details

Gene
IGDCC3
Gene Name
immunoglobulin superfamily DCC subclass member 3
There are no transcripts associated with this gene.

Gene
IGDCC4
Gene Name
immunoglobulin superfamily DCC subclass member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020962.2 3227 Missense Mutation AGC,AGG S1070R NP_066013.1
XM_011521845.2 3227 Missense Mutation AGC,AGG S1083R XP_011520147.1
XM_011521846.2 3227 Missense Mutation AGC,AGG S1071R XP_011520148.1
XM_011521847.1 3227 Missense Mutation AGC,AGG S809R XP_011520149.1
XM_017022448.1 3227 Missense Mutation AGC,AGG S1084R XP_016877937.1
XM_017022449.1 3227 Intron XP_016877938.1

View Full Product Details