Product Details

SNP ID
rs187004394
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:84479344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAGTCTAAAACGGAAAGCCGC[G/T]GCCGAAGAACTTCAGGAGGCTGCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPF1 PubMed Links

Gene Details

Gene
RPF1
Gene Name
ribosome production factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025065.6 108 Silent Mutation GCG,GCT A21A NP_079341.2

View Full Product Details