Product Details

SNP ID
rs186998770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55529935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCGGTCCCGGGGCTGGCCCGAC[A/G]CCTGCCAGATGAGGAAGTCCTCGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SBK2 PubMed Links

Gene Details

Gene
SBK2
Gene Name
SH3 domain binding kinase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101401.2 860 Missense Mutation NP_001094871.2
XM_006723327.3 860 Missense Mutation XP_006723390.1
XM_011527227.2 860 Intron XP_011525529.1

View Full Product Details