Product Details

SNP ID
rs186779400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:48029377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTAGGGCCGCGCCGCCGCCAC[A/G]TTCTCGGCCAGCGCGAAGGCCAGGA
Phenotype
MIM: 616591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB7C PubMed Links

Gene Details

Gene
ZBTB7C
Gene Name
zinc finger and BTB domain containing 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039360.2 2456 Silent Mutation NP_001034449.1
NM_001318841.1 2456 Silent Mutation NP_001305770.1
XM_005258229.4 2456 Silent Mutation XP_005258286.1
XM_011525861.2 2456 Silent Mutation XP_011524163.1
XM_011525863.2 2456 Silent Mutation XP_011524165.1
XM_011525864.2 2456 Missense Mutation XP_011524166.1
XM_011525865.2 2456 Silent Mutation XP_011524167.1
XM_011525866.2 2456 Silent Mutation XP_011524168.1
XM_011525867.2 2456 Silent Mutation XP_011524169.1
XM_011525869.2 2456 Silent Mutation XP_011524171.1
XM_011525870.2 2456 Silent Mutation XP_011524172.1
XM_011525871.2 2456 Silent Mutation XP_011524173.1
XM_017025605.1 2456 Silent Mutation XP_016881094.1
XM_017025606.1 2456 Silent Mutation XP_016881095.1
XM_017025607.1 2456 Silent Mutation XP_016881096.1
XM_017025608.1 2456 Silent Mutation XP_016881097.1
XM_017025609.1 2456 Silent Mutation XP_016881098.1

View Full Product Details