Product Details

SNP ID
rs187608701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44568915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATCCACTCACCTTAAGCTTCTC[A/C]GAATGCAGAAGCTCTTCCTTGATCT
Phenotype
MIM: 608023 MIM: 612038
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DDX56 PubMed Links

Gene Details

Gene
DDX56
Gene Name
DEAD-box helicase 56
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257189.1 1894 Silent Mutation TCG,TCT S417S NP_001244118.1
NM_019082.3 1894 Silent Mutation TCG,TCT S457S NP_061955.1
Gene
TMED4
Gene Name
transmembrane p24 trafficking protein 4
There are no transcripts associated with this gene.

View Full Product Details