Product Details

SNP ID
rs187611065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37024485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCGGAGTACGCAGGCCGCCTGGG[C/T]GTGGGCGCGGCCACCCACGTCGTGA
Phenotype
MIM: 602496 MIM: 180370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPST PubMed Links

Gene Details

Gene
MPST
Gene Name
mercaptopyruvate sulfurtransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013436.2 427 Silent Mutation GGC,GGT G90G NP_001013454.1
NM_001130517.2 427 Silent Mutation GGC,GGT G90G NP_001123989.1
NM_021126.5 427 Silent Mutation GGC,GGT G110G NP_066949.2
XM_005261610.3 427 Silent Mutation GGC,GGT G90G XP_005261667.1
XM_011530195.1 427 Silent Mutation GGC,GGT G110G XP_011528497.1
XM_011530196.1 427 Silent Mutation GGC,GGT G90G XP_011528498.1
Gene
TST
Gene Name
thiosulfate sulfurtransferase
There are no transcripts associated with this gene.

View Full Product Details