Product Details

SNP ID
rs187627657
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88521666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATAGGCAAGAGCAGGCTGTAAA[A/G]GCAAAGGCTGGCTGTGCTAGTGCAG
Phenotype
MIM: 610662
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929134 PubMed Links

Gene Details

Gene
LOC101929134
Gene Name
uncharacterized LOC101929134
There are no transcripts associated with this gene.

Gene
PIGY
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Y
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042616.2 668 Missense Mutation CTT,TTT L42F NP_001036081.1
Gene
PYURF
Gene Name
PIGY upstream reading frame
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032906.4 668 UTR 3 NP_116295.1

View Full Product Details