Product Details

SNP ID
rs187623535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:100990575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATATTCTAGCCATCAAATCAAT[C/T]CTATACTCACTGAGAGGCTCAAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL13A PubMed Links

Gene Details

Gene
ARL13A
Gene Name
ADP ribosylation factor like GTPase 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162491.1 1797 Missense Mutation TCC,TTC S253F NP_001155963.1
XM_011530950.2 1797 UTR 3 XP_011529252.1
XM_011530951.2 1797 Intron XP_011529253.1
Gene
TRMT2B
Gene Name
tRNA methyltransferase 2 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167970.1 1797 Intron NP_001161442.1
NM_001167971.1 1797 Intron NP_001161443.1
NM_001167972.1 1797 Intron NP_001161444.1
NM_024917.5 1797 Intron NP_079193.2
XM_005262195.2 1797 Intron XP_005262252.1
XM_005262196.2 1797 Intron XP_005262253.1
XM_005262201.2 1797 Intron XP_005262258.1
XM_006724705.2 1797 Intron XP_006724768.1
XM_011531042.2 1797 Intron XP_011529344.1
XM_011531044.2 1797 Intron XP_011529346.1
XM_017029858.1 1797 Intron XP_016885347.1
XM_017029859.1 1797 Intron XP_016885348.1
XM_017029860.1 1797 Intron XP_016885349.1
XM_017029861.1 1797 Intron XP_016885350.1
XM_017029862.1 1797 Intron XP_016885351.1
XM_017029863.1 1797 Intron XP_016885352.1
XM_017029864.1 1797 Intron XP_016885353.1
XM_017029865.1 1797 Intron XP_016885354.1
XM_017029866.1 1797 Intron XP_016885355.1
XM_017029867.1 1797 Intron XP_016885356.1
XM_017029868.1 1797 Intron XP_016885357.1
XM_017029869.1 1797 Intron XP_016885358.1
XM_017029870.1 1797 Intron XP_016885359.1
XM_017029871.1 1797 Intron XP_016885360.1
XM_017029872.1 1797 Intron XP_016885361.1
XM_017029873.1 1797 Intron XP_016885362.1
XM_017029874.1 1797 Intron XP_016885363.1
XM_017029875.1 1797 Intron XP_016885364.1

View Full Product Details