Product Details
- SNP ID
-
rs187660587
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:103563113 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCGGCGCCGAGCGCAGGGATACG[A/G]CGCCCAGCGGGGTAAGCAGGGGCCT
- Phenotype
-
MIM: 616003
MIM: 603885
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APOPT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2274267] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APOPT1
- Gene Name
- apoptogenic 1, mitochondrial
- Gene
- BAG5
- Gene Name
- BCL2 associated athanogene 5
View Full Product Details