Product Details

SNP ID
rs187785472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144095764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGTGCTGGTCTCCTACTTGCA[C/T]ATCCTGGTGGCCATCTTGAGGATCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2A12 PubMed Links

Gene Details

Gene
OR2A12
Gene Name
olfactory receptor family 2 subfamily A member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004135.1 657 Silent Mutation CAC,CAT H219H NP_001004135.1

View Full Product Details