Product Details

SNP ID
rs187710744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112275878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGTGAGTTTTTACCACTTCGTCA[A/C]CTGTCGGCGGCGGCCGGGAGCAGGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZC3H6 PubMed Links

Gene Details

Gene
ZC3H6
Gene Name
zinc finger CCCH-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198581.2 285 UTR 5 NP_940983.2
XM_006712519.2 285 UTR 5 XP_006712582.1

View Full Product Details