Product Details

SNP ID
rs187441827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120875329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAGCCAATGCCCGCCGCCGGGTA[C/G]CGACGGGTGGCCACCGCCAAGTCGA
Phenotype
MIM: 300790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CT47A12 PubMed Links

Gene Details

Gene
CT47A12
Gene Name
cancer/testis antigen family 47, member A12
There are no transcripts associated with this gene.

Gene
CT47B1
Gene Name
cancer/testis antigen family 47, member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145718.1 559 Intron NP_001139190.1
XM_017029734.1 559 Silent Mutation CGC,CGG R114R XP_016885223.1

View Full Product Details