Product Details

SNP ID
rs187397797
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69433556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCTCCTTCACCACAGCTGCA[C/T]AGGATTTATTATATGGATCATTATT
Phenotype
MIM: 611964
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CYB5B PubMed Links

Gene Details

Gene
CYB5B
Gene Name
cytochrome b5 type B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030579.2 Intron NP_085056.2

View Full Product Details