Product Details

SNP ID
rs187427561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132538929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTTTATACCCAATGTCGCCATG[A/G]TGTTTATATACAGTAAGATATTTTT
Phenotype
MIM: 608282
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAAR9 PubMed Links

Gene Details

Gene
TAAR9
Gene Name
trace amine associated receptor 9 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175057.3 642 Missense Mutation ATG,GTG M214V NP_778227.3

View Full Product Details