Product Details
- SNP ID
-
hCV183100805
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62464963 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACTTGAACTCCAAGTGGCCGGGG[C/G]CAAGAGAGTCATCCTCCTGGCCAGA
- Phenotype
-
MIM: 611496
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GATA5
PubMed Links
Gene Details
- Gene
- GATA5
- Gene Name
- GATA binding protein 5
There are no transcripts associated with this gene.
View Full Product Details