Product Details

SNP ID
rs187492563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10280498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAATTAAATTCCTTCTGGCCTC[G/T]TCAATGTAGGGCTTGTCTGCAGGTG
Phenotype
MIM: 613379
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CMBL PubMed Links

Gene Details

Gene
CMBL
Gene Name
carboxymethylenebutenolidase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138809.3 1144 Silent Mutation NP_620164.1
XM_017009034.1 1144 Silent Mutation XP_016864523.1
XM_017009035.1 1144 Silent Mutation XP_016864524.1

View Full Product Details