Product Details
- SNP ID
-
rs187574172
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:223393549 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACCCCCGCGGCCCGGCGGAGAAC[C/G]AGAGTCCACGCAGCGGCTGGGCGGG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC185
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111306873] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC185
- Gene Name
- coiled-coil domain containing 185
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152610.2 |
177 |
Missense Mutation |
CCA,CGA |
P25R |
NP_689823.2 |
View Full Product Details