Product Details

SNP ID
rs187303852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:103108929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGCACAGGATCTTGTAAATTCC[A/G]TTCCTTGCATTGAGAGGGTGTAATT
Phenotype
MIM: 117143
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BDH2 PubMed Links

Gene Details

Gene
BDH2
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
There are no transcripts associated with this gene.

Gene
CENPE
Gene Name
centromere protein E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286734.1 7975 Missense Mutation CGG,TGG R2508W NP_001273663.1
NM_001813.2 7975 Missense Mutation CGG,TGG R2629W NP_001804.2
XM_011531544.2 7975 Missense Mutation CGG,TGG R2604W XP_011529846.1
XM_011531545.2 7975 Missense Mutation CGG,TGG R2576W XP_011529847.1
XM_011531546.2 7975 Missense Mutation CGG,TGG R2561W XP_011529848.1
XM_011531547.2 7975 Missense Mutation CGG,TGG R2533W XP_011529849.1
XM_011531548.2 7975 Missense Mutation CGG,TGG R2532W XP_011529850.1
XM_011531549.2 7975 Missense Mutation CGG,TGG R2481W XP_011529851.1
XM_017007659.1 7975 Missense Mutation CGG,TGG R2479W XP_016863148.1

View Full Product Details