Product Details

SNP ID
rs187261074
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39665905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTTCACCCCTGCCAAGATGGG[C/T]GCCACCAAGGAGGAGCGTGAGGACA
Phenotype
MIM: 611801 MIM: 171190 MIM: 607048 MIM: 604488
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGAP3 PubMed Links

Gene Details

Gene
PGAP3
Gene Name
post-GPI attachment to proteins 3
There are no transcripts associated with this gene.

Gene
PNMT
Gene Name
phenylethanolamine N-methyltransferase
There are no transcripts associated with this gene.

Gene
STARD3
Gene Name
StAR related lipid transfer domain containing 3
There are no transcripts associated with this gene.

Gene
TCAP
Gene Name
titin-cap
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003673.3 314 Silent Mutation GGC,GGT G100G NP_003664.1

View Full Product Details