Product Details
- SNP ID
-
rs187291577
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:138424439 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGGGGCCGGAAAAGGACAGACTC[C/T]GAGGCATCAAGCCTTCGTTCTTGGC
- Phenotype
-
MIM: 605825
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HEBP2
PubMed Links
Gene Details
- Gene
- HEBP2
- Gene Name
- heme binding protein 2
There are no transcripts associated with this gene.
- Gene
- NHSL1
- Gene Name
- NHS like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144060.1 |
6801 |
Missense Mutation |
CAG,CGG |
Q1488R |
NP_001137532.1 |
NM_020464.1 |
6801 |
Missense Mutation |
CAG,CGG |
Q1492R |
NP_065197.1 |
XM_005267062.4 |
6801 |
Missense Mutation |
CAG,CGG |
Q1536R |
XP_005267119.1 |
XM_011535965.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1554R |
XP_011534267.1 |
XM_011535966.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1554R |
XP_011534268.1 |
XM_011535967.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1554R |
XP_011534269.1 |
XM_011535968.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1510R |
XP_011534270.1 |
XM_011535969.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1507R |
XP_011534271.1 |
XM_011535970.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1506R |
XP_011534272.1 |
XM_011535971.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1500R |
XP_011534273.1 |
XM_011535972.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1492R |
XP_011534274.1 |
XM_011535973.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1489R |
XP_011534275.1 |
XM_011535974.2 |
6801 |
Missense Mutation |
CAG,CGG |
Q1489R |
XP_011534276.1 |
XM_011535976.1 |
6801 |
Missense Mutation |
CAG,CGG |
Q1430R |
XP_011534278.1 |
XM_017011086.1 |
6801 |
Missense Mutation |
CAG,CGG |
Q1554R |
XP_016866575.1 |
XM_017011087.1 |
6801 |
Missense Mutation |
CAG,CGG |
Q1519R |
XP_016866576.1 |
XM_017011088.1 |
6801 |
Missense Mutation |
CAG,CGG |
Q1504R |
XP_016866577.1 |
XM_017011089.1 |
6801 |
Missense Mutation |
CAG,CGG |
Q1474R |
XP_016866578.1 |
View Full Product Details