Product Details

SNP ID
rs187288596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58084925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCGGCCGCCATGGCTGCTGTG[A/G]TGCTCCATCAGGTGCGAGAGCCACG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN18 PubMed Links

Gene Details

Gene
ZSCAN18
Gene Name
zinc finger and SCAN domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145542.1 1491 Silent Mutation NP_001139014.1
NM_001145543.1 1491 Silent Mutation NP_001139015.1
NM_001145544.1 1491 Silent Mutation NP_001139016.1
NM_023926.4 1491 Silent Mutation NP_076415.3
XM_005259174.4 1491 Silent Mutation XP_005259231.1
XM_006723335.2 1491 Silent Mutation XP_006723398.1
XM_011527237.2 1491 Silent Mutation XP_011525539.1
XM_011527238.1 1491 Silent Mutation XP_011525540.1
XM_011527239.2 1491 Silent Mutation XP_011525541.1
XM_017027169.1 1491 Silent Mutation XP_016882658.1
XM_017027170.1 1491 Silent Mutation XP_016882659.1
XM_017027171.1 1491 Missense Mutation XP_016882660.1

View Full Product Details