Product Details

SNP ID
rs187325799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49684808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACGCTGTATGTGACGGCCATC[A/G]AGGACCGGCAGTACAAAGACTACAA
Phenotype
MIM: 608846 MIM: 602950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADM5 PubMed Links

Gene Details

Gene
ADM5
Gene Name
adrenomedullin 5 (putative)
There are no transcripts associated with this gene.

Gene
CPT1C
Gene Name
carnitine palmitoyltransferase 1C
There are no transcripts associated with this gene.

Gene
MIR5088
Gene Name
microRNA 5088
There are no transcripts associated with this gene.

Gene
PRMT1
Gene Name
protein arginine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207042.2 727 Intron NP_001193971.1
NM_001536.5 727 Missense Mutation AAG,GAG K204E NP_001527.3
NM_198318.4 727 Missense Mutation AAG,GAG K186E NP_938074.2
XM_005258842.1 727 Missense Mutation AAG,GAG K175E XP_005258899.1
XM_017026734.1 727 Missense Mutation AAG,GAG K180E XP_016882223.1
XM_017026735.1 727 Missense Mutation AAG,GAG K180E XP_016882224.1
XM_017026736.1 727 Missense Mutation AAG,GAG K175E XP_016882225.1

View Full Product Details