Product Details

SNP ID
rs187905695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51078900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGGATAGGCCTTCTGGCACACC[C/T]CATCCGGGGAGATGTTGATGTTCAC
Phenotype
MIM: 606135
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK14 PubMed Links

Gene Details

Gene
KLK14
Gene Name
kallikrein related peptidase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311182.1 750 Missense Mutation GAG,GGG E189G NP_001298111.1
NM_022046.5 750 Missense Mutation GAG,GGG E189G NP_071329.2
XM_017026820.1 750 UTR 3 XP_016882309.1

View Full Product Details