Product Details
- SNP ID
-
rs188195414
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:41714045 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCAGCTGAAGCTGGAGCAGACTAT[C/T]CGTTGCCTGCTGGATGAGTGCCACA
- Phenotype
-
MIM: 608797
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MEI1
PubMed Links
Gene Details
- Gene
- MEI1
- Gene Name
- meiotic double-stranded break formation protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152513.3 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
NP_689726.3 |
XM_011529935.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528237.1 |
XM_011529936.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528238.1 |
XM_011529937.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528239.1 |
XM_011529938.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528240.1 |
XM_011529939.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528241.1 |
XM_011529940.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528242.1 |
XM_011529941.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528243.1 |
XM_011529942.2 |
427 |
Silent Mutation |
ATC,ATT |
I67I |
XP_011528244.1 |
XM_011529943.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528245.1 |
XM_011529944.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528246.1 |
XM_011529945.2 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528247.1 |
XM_011529946.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528248.1 |
XM_011529947.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528249.1 |
XM_011529948.2 |
427 |
Intron |
|
|
XP_011528250.1 |
XM_011529949.1 |
427 |
UTR 5 |
|
|
XP_011528251.1 |
XM_011529950.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528252.1 |
XM_011529952.1 |
427 |
Intron |
|
|
XP_011528254.1 |
XM_011529954.1 |
427 |
Intron |
|
|
XP_011528256.1 |
XM_011529955.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528257.1 |
XM_011529956.2 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_011528258.1 |
XM_017028633.1 |
427 |
Silent Mutation |
ATC,ATT |
I131I |
XP_016884122.1 |
XM_017028635.1 |
427 |
Intron |
|
|
XP_016884124.1 |
View Full Product Details