Product Details

SNP ID
rs188182718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41938660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCAACTTCTGGAGGCAGCAGAA[A/G]CCGATCTACGCCAGGGAGCGGGACC
Phenotype
MIM: 108728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACLY PubMed Links

Gene Details

Gene
ACLY
Gene Name
ATP citrate lyase
There are no transcripts associated with this gene.

Gene
TTC25
Gene Name
tetratricopeptide repeat domain 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031421.3 830 Silent Mutation AAA,AAG K243K NP_113609.1
XM_011525323.2 830 Silent Mutation AAA,AAG K80K XP_011523625.1
XM_011525324.2 830 Silent Mutation AAA,AAG K243K XP_011523626.1

View Full Product Details